Learn More
CleanWear™ Tyvek™ Disposable Coveralls with Elastic Ankles and Wrists
Tough coverall protects skin from wet and dry particulate matter
Manufacturer:  Cleanwear USA 11548XL
 View more versions of this product
Catalog No. 18-892-411
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper front
Elastic wrist and ankle, collar
White
Yes
Tyvek™ Spunbond Olefin
Serged
8X-Large
Excellent durability, breathability and barrier protection and ensures wearer comfort by allowing air and moisture vapor to pass through while maintaining a virtually impermeable particulate barrier