Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Clear Caps

Use with empty FastPrep™ tubes. MP Biomedicals Clear Caps are screw scaps with an O-ring for use with 2mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix tubes for prepare samples for homogenization

$138.30 - $341.50

Specifications

Storage Requirements Room Temperature (15 to 30°C)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP115068002
View Documents
MP Biomedicals™
115068002
500 Caps
Each for $138.30
Add to cart
 
MP115068005
View Documents
MP Biomedicals™
115068005
1000 Caps
Each for $341.50
Add to cart
 
Description

Description

Clear Caps fit empty FastPrep™ tubes and are used in FastPrep systems. Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.

Specifications

Specifications

Room Temperature (15 to 30°C)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit