Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Clear Double-Sided Laminates

Double-sided/Industrial use

$118.00 - $202.50

Specifications

Product Type Double-Sided Laminate
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Size (Metric) Price Quantity & Availability  
19-104-507
View Documents
Brady™
20589
8.6 in. x 50 ft. 21.8 cm x 15.24m Each for $118.00
Add to cart
 
19-104-508
View Documents
Brady™
20602
8.6 in. x 100 ft. 21.8 cm x 30.4m Each for $202.50
Add to cart
 
Description

Description

  • Laminates documents 8.6 in. wide and up to 100 ft. long
  • Constructed from tough polyester
  • Available in sizes 8.6 in. x 50 ft. and 8.6 in. x 100 ft.
Specifications

Specifications

Double-Sided Laminate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit