Fisherbrand™ Graduated Glass Bottle Beakers without Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers without Cap can also be used for storing industrial laboratory chemicals and reagents.

 View more versions of this product

Catalog No. 02-912-257A

  Check Availability
Add to Cart

Includes: Clear Graduated AC Round Bottle

    Specifications

    Capacity (Metric) 240mL
    Neck Finish 58-400
    Shape Round
    Capacity (English) 8 oz.
    Product Type Graduated Bottle
    Color Clear
    Material Glass
    Mouth Wide
    Quantity 96/Cs.

    • Graduated in both milliliters and ounces
    • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
    • Wide mouth facilitates easy mixing, handling, and transfer of contents

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit