Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap

Environmentally sensitive bottles help eliminate waste. Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap ensures product integrity for long term storage.

 View more versions of this product

Catalog No. 02-911-458A

  Check Availability
Add to Cart

Includes: Polypropylene PE Foam/PTFE lined cap

    Specifications

    Product Type Graduated Bottle
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Color Clear
    Graduated Yes
    Liner Material PE Foam/PTFE
    Mouth Wide
    Neck Finish 33-400
    Quantity 48/Cs.
    Shape Round
    Capacity (Metric) 30mL
    Capacity (English) 1 oz.
    Material Glass

    Clear Graduated Boston Round Bottles
    • Feature a narrow screw neck opening, curved shoulders, and a round body
    • Glass offers minimal chemical interference with contents
    • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
     Polypropylene PE Foam/PTFE Lined Caps
    • Offer good impact strength, thermal resistance and pliability
    • Autoclavable
    • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
    • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit