Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.

 View more versions of this product

Catalog No. 02-911-457A



Includes: Polypropylene Pulp/Vinyl lined cap

Description

Description

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications

Specifications

480mL
Polypropylene
GlassþGlass
Bottle, GraduatedþGraduated Medium Round Bottle
Pulp/Vinyl
70-400
Screw
16 oz.
ClearþClear
24/Cs.
White
Wide
Round
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit