Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand Clear Graduated Glass Bottles with White Polypropylene Caps
Fisherbrand Clear Graduated Glass Bottles with White Polypropylene Caps feature a wide mouth for easy removal of contents.
Description
- Graduated in both milliliters and ounces
- Clear glass bottles are designed with a slight shoulder for easy pouring and handling
- Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
60mL | |
Polypropylene | |
Clear | |
Type III Glass | |
Yes | |
Wide Mouth |
2 oz | |
Pulp/Vinyl | |
Yes | |
48 | |
White | |
38-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title