Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clear Plastic Terrarium/Aquarium Products for Science Education

Use as an aquarium, terrarium or animal cage.

Manufacturer:  Nasco SB15079DIM

 View more versions of this product

Catalog No. S17740


Add to cart

Description

Description

Clear plastic tanks are ideally suited for fish, frogs, chameleons, land hermit crabs, insects, spiders, gerbils, mice and terrarium plants.

  • Flexible plastic material resists cracking and breakage
  • Tanks conveniently nest inside each other for storage
  • Thickness: 80mil

Order Info

Lid is included with S17740; for S17740-1, it must be purchased separately.

Specifications

Specifications

1.5 gal.
Use as an aquarium, terrarium, or animal cage for Fish, Frogs, Chameleons, Land Hermit Crabs, insects, Spiders, Gerbils, Mice and terrarium plants
11 in.
Plastic
Education
5.7L
Lid
27.9cm
Flex tank and domed cover with dial ventilator
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit