Learn More
Thermo Scientific™ I-Chem™ Clear VOA Glass Vials with Closed-Top Cap
Use these clear glass vials with closed-top caps for EPA volatile organic analysis (VOA) when a pierceable septa closure is not needed.
$173.80 - $345.50
Specifications
No. per Case 72
Closure Size 24-414
Diameter (English) Outer 1.1 in.
Diameter (Metric) Outer 28 mm
Color Clear
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Level Description Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Level Description Price Quantity & Availability  
05-719-111
View Documents
Thermo Scientific™
C1260020
20 mL Unprocessed I-Chem 20mL Vial clear closed top unprocessed
Case of 72 for $173.80
Edge
Add to cart
05-719-120
View Documents
Thermo Scientific™
C1360040
40 mL Unprocessed I-Chem 40mL Vial clear closed top unprocessed
Case of 72 for $207.00
Edge
Add to cart
05-719-110
View Documents
Thermo Scientific™
C2260020
20 mL Processed I-Chem 20mL Vial clear closed top processed
Case of 72 for $268.00
Add to cart
05-719-119
View Documents
Thermo Scientific™
C2360040
40 mL Processed I-Chem 40mL Vial clear closed top processed
Case of 72 for $299.50
Edge
Add to cart
05-719-109
View Documents
Thermo Scientific™
C3260020
20 mL Certified I-Chem 20mL Vial clear closed top certified
Case of 72 for $317.50
Add to cart
05-719-118
View Documents
Thermo Scientific™
C3360040
40 mL Certified I-Chem 40mL Vial clear closed top certified
Case of 72 for $345.50
Edge
Add to cart
Description
Description
Thermo Scientific™ Clear VOA Vials with Closed Cap are designed for EPA volatile organic analysis (VOA). Closed-top vials for VOA sampling when a pierceable septa closure is not needed. Available Certified to meet EPA Performance Based Specifications for Volatile Organic Analysis, Processed the same way as the certified vials but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Clear vial designed for volatile organic analysis (VOA)
  • Choose from 20mL or 40mL sizes
  • Closed-top white polypropylene cap with PTFE-faced silicone liner bonded to the cap for a secure seal
  • Directly compatible with automation
  • Closures assembled to vials
Includes:
Certified vials come with the following features:
  • Certified to meet or exceed EPA performance-based standards for volatile organic analysis
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each vial individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Unique Field to Lab carton conveniently converts to a vial organizer tray for convenient lab use. Chipboard divider packaging to cushion and help protect vials from breakage; overpack required for shipping
Processed vials are processed in the same manner as Certified vials and come with all the features of
Certified vials, but vials are not barcoded, and CofA is not included.
Unprocessed vials are economically packaged glass-to-glass in a shrink-wrapped tray and are ready for your own in-house cleaning and certification process.
Specifications
Specifications
72
1.1 in.
Clear
B267-2414UBB
24-414
28 mm
Glass (Type 1)