Learn More
MSA™ ClearLITE™ Plus Face Shield
Fit easily on MSA face shield frames
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
View More Specs
Alert:
  • Visors should always be worn with primary eye protection such as Z87.1-2003 high-impact resistant spectacles or goggles
  • Do not use these visors for welding, elevated temperature applications, or for protection against other infrared radiation sources unless it is approved for such applications
  • Do not use for sports, simulated war games, paintball games, or similar activities
  • Inspect visor frequently and replace immediately if worn, scratched, or damaged
  • Do not use near open flame
Products 2
Catalog Number Mfr. No. Length (English) Lens Material Lens Tint Shape Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Lens Material Lens Tint Shape Price Quantity & Availability  
19-079846
View Documents
MSA™
488132
7.88 in. Polycarbonate Clear Formed This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
18-320-730
View Documents
MSA™
10017934
7.75 in. Polycarbonate Clear Flat This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Meets ANSI Z87.1-2003 standards.