Learn More
CLEC-1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0187-8K42, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212192AF350
Catalog No. NP212192A30
Add to cart
Description
Description
CLEC-1 Monoclonal antibody specifically detects CLEC-1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CLEC-1
MM0187-8K42
Western Blot
CLEC-1, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CLEC1A
Human CLEC1A
0.1 mL
Primary
51267