Learn More
CLEC-2/CLEC-1B Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0188-8J24, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212193AF532
Catalog No. NP212193A32
Add to cart
Description
Description
CLEC-2/CLEC-1B Monoclonal antibody specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Western Blot.
Specifications
Specifications
CLEC-2/CLEC-1B
MM0188-8J24
Western Blot
1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CLEC1B
Human CLEC1B
0.1 mL
Primary
51266