Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC-2 (CLEC1B) Mouse anti-Human, Brilliant Violet 510, Clone: 219133, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 748139
Description
CLEC-2 (CLEC1B) Monoclonal antibody specifically detects CLEC-2 (CLEC1B) in Human samples. It is validated for Flow Cytometry.Specifications
CLEC-2 (CLEC1B) | |
219133 | |
Brilliant Violet 510 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal | |
Human |
Flow Cytometry | |
0.2 mg/mL | |
Conjugated, Purified | |
IgG2a, κ | |
RUO | |
Primary | |
CLEC-2 (CLEC1B) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title