Learn More
CLEC11A Mouse anti-Human, Clone: 3E1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524412
Catalog No. PIMA524412
Add to cart
Description
Description
Peptide Sequence: PADRQQMEAL TRYLRAALAP YNWPVWLGVH DRRAEGLYLF ENGQRVSFFA WHRSPRPELG AQPSASPHPL SPDQPNGGTL ENCVAQASDD GSWWDHDCQR RLYYVCEFPF
Stimulates the proliferation and differentiation of hematopoietic precursor cells from various lineages, including erythrocytes, lymphocytes, granulocytes and macrophages. Acts synergistically with other cytokines, including IL-3, GCSF, GMCSF and FLT3 ligand. Suppresses SCF-stimulated erythrocyte proliferation.
Specifications
Specifications
CLEC11A
Monoclonal
Unconjugated
CLEC11A
Liquid
CLEC11A
CLEC11A (NP_002966, 214 a.a. ∽ 323 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA
3E1
PBS with no preservative; pH 7.4
Q9Y240
C-type lectin domain family 11 member A, C-type lectin superfamily member 3, Lymphocyte secreted C-type lectin, Stem cell growth factor, p47, CLEC11A, CLECSF3, LSLCL, SCGF
Mouse
IgG2b, lambda
100 μg
-20° C
Primary
6320