Learn More
CLEC14A Mouse anti-Human, Alexa Fluor 647, Clone: 743940, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7436R100UG
Catalog No. FAB7436R100
Add to cart
This item is not returnable. View return policy
Description
Description
CLEC14A Monoclonal antibody specifically detects CLEC14A in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CLEC14A
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q86T13
CLEC14A
Mouse myeloma cell line NS0-derived recombinant human CLEC14A Glu22-Ala397 Accession # Q86T13
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human CLEC14A in direct ELISA.
Flow Cytometry
743940
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGF R5, EGFR5, EGFR-5, Epidermal growth factor receptor 5
Mouse
IgG2b
RUO
Primary
161198
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)