Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550210

Catalog No. PA550210


Add to cart

Description

Description

The antibody detects endogenous levels of total CLEC2 / CLEC1B protein.

Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D , that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells .
Specifications

Specifications

CLEC1B
Polyclonal
Unconjugated
CLEC1B
Liquid
CLEC1B
Full length fusion protein of Human CLEC1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9P126
CLEC2
Rabbit
IgG
100 μL
-20°C
Primary
51266
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit