Learn More
CLEC3B/Tetranectin Mouse anti-Bovine, Janelia Fluor 549, Clone: 5B7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105142JF549
Catalog No. NBP15142549
Add to cart
Description
Description
CLEC3B/Tetranectin Monoclonal antibody specifically detects CLEC3B/Tetranectin in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CLEC3B/Tetranectin
5B7
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CLEC3B
Tetranectin purified from human citrate plasma (3) and coupled to PPD. Boosted before fusion with recombinant tetranectin produced in E. coli.
Protein A or G purified
Cancer
Primary
7123
Bovine, Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Mouse
IgG2a Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Specific for amino acids 17-181 of human tetranectin monomer