Learn More
CLEC3B/Tetranectin Mouse anti-Human, Alexa Fluor 405, Clone: MM0639-7G16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP232726AF405
Catalog No. NB232726405
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CLEC3B/Tetranectin Monoclonal antibody specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
CLEC3B/Tetranectin
MM0639-7G16
Western Blot, ELISA
Purified
CLEC3B
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human Tetranectin
Protein G purified
Cancer
Primary
7123
Western Blot, ELISA
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Human