Learn More
CLEC3B/Tetranectin Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0639-7G16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232726AF594
Catalog No. NP232726A94
Add to cart
Description
Description
CLEC3B/Tetranectin Monoclonal antibody specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
CLEC3B/Tetranectin
MM0639-7G16
Western Blot, ELISA
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CLEC3B
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human Tetranectin
0.1 mL
Primary
7123