Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLEC4C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109550
Description
Immunogen sequence: NFMYSKTVKR LSKLREYQQY HPSLTCVMEG KDIEDWSCCP TPWTSFQSSC YFISTGMQSW TKSQKNCSVM GADLVVINTR EEQDFIIQNL KRNSSYFLGL SDPGGRRHWQ WVDQTPYNEN VTFWHSGEPN NLDERCAIIN FRSSEEWGWN DIHCHVPQKS ICKMKKIYI.
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
CLEC4C | |
Polyclonal | |
Unconjugated | |
CLEC4C | |
BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
170482 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.56 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8WTT0 | |
CLEC4C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 45-213 of human CLEC4C (NP_569708.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction