Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC4C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 179411AP150UL
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signaling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
CLEC4C | |
Polyclonal | |
Unconjugated | |
CLEC4C | |
BDCA 2, BDCA2, Blood dendritic cell antigen 2, CD303, CLEC4C, CLECSF11, CLECSF7, Dendritic lectin, DLEC, HECL, PRO34150 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170482 | |
-20°C | |
Liquid |
ELISA | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WTT0 | |
CLEC4C | |
CLEC4C Fusion Protein Ag12363 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction