Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC4D/CLECSF8 Antibody (413512), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2806MFV610
Description
CLEC4D/CLECSF8 Monoclonal antibody specifically detects CLEC4D/CLECSF8 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| CLEC4D/CLECSF8 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 338339 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 413512 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8, Gly52-Asn215, Accession # Q8WXI8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction