Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CLEC4D/CLECSF8 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 24 pg/mL |
---|---|
Assay Sensitivity | 23.44-1500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010315
|
Novus Biologicals™
NBP27963815PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human CLEC4D/CLECSF8.Specifications
24 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Human CLEC4D/CLECSF8. | |
Sandwich ELISA | |
338339 | |
Rabbit/Mouse | |
RUO | |
CLEC4D/CLECSF8 |
23.44-1500 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL | |
CLEC4D | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title