Learn More
CLEC4D Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549700
Catalog No. PA549700
Add to cart
Description
Description
The antibody detects endogenous level of total CLEC4D / CLECSF8 protein.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
Specifications
CLEC4D
Polyclonal
Unconjugated
CLEC4D
Liquid
CLEC4D
Fusion protein corresponding to a region derived from 39-215 amino acids of human C-type lectin domain family 4, member D
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8WXI8
CLECSF8, MCL
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
338339