Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4E Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271793JF549

Catalog No. N271793J549


Add to cart

Description

Description

CLEC4E Monoclonal antibody specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

CLEC4E
OTI2A8
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
CLEC4E
Full length recombinant protein of human CLEC4E (NP_055173) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9
Mouse
IgG1
0.1 mL
Primary
26253.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit