Learn More
CLEC4E Rabbit anti-Human, Alexa Fluor 405, Clone: 2455C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB8995V100UG
Catalog No. FAB8995V100
Add to cart
This item is not returnable. View return policy
Description
Description
CLEC4E Monoclonal antibody specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CLEC4E
2455C
Flow Cytometry 0.25-1 ug/10^6 cells
Q9ULY5
CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, MINCLE, MINCLEC-type lectin superfamily member 9
Chinese Hamster Ovary cell line, CHO-derived recombinant human CLEC4E, Arg41-Leu219, Accession # Q9ULY5
Protein A or G purified from cell culture supernatant
RUO
Primary
26253
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rabbit
IgG
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human