Learn More
CLEC4G Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181731AP
Catalog No. 50-173-2029
Add to cart
Description
Description
This gene encodes a glycan-binding receptor and member of the C-type lectin family which plays a role in the T-cell immune response. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CLEC4G
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXB4
CLEC4G, LP2698, LSECtin, UNQ431
Rabbit
IgG
150 μL
-20°C
Primary
339390
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CLEC4G
Liquid
CLEC4G
CLEC4G Fusion Protein Ag12837
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human