Learn More
CLEC4M Mouse anti-Human, Clone: OTI10C3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810055
Catalog No. 50-166-9863
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CLEC4M (C-Type Lectin Domain Family 4 Member M) is a Protein Coding gene. Diseases associated with CLEC4M include Japanese Encephalitis and Severe Acute Respiratory Syndrome. Among its related pathways are C-type lectin receptor signaling pathway and Phagosome. Gene Ontology (GO) annotations related to this gene include calcium-dependent protein binding. An important paralog of this gene is CD209.
Specifications
Specifications
CLEC4M
Monoclonal
Unconjugated
CLEC4M
Lyophilized
CLEC4M
Human recombit protein fragment corresponding to amino acids 74-329 of human CLEC4M produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI10C3
PBS with 8% trehalose and no preservative; pH 7.3
Q9H2X3
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10332