Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4M Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen CLEC4M
Concentration 0.38 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-3677
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
220031AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

CLEC4M (C-Type Lectin Domain Family 4 Member M) is a Protein Coding gene. Diseases associated with CLEC4M includeJapanese EncephalitisandSevere Acute Respiratory Syndrome. Among its related pathways areC-type lectin receptor signaling pathwayandPhagosome. Gene Ontology (GO) annotations related to this gene includecalcium-dependent protein binding. An important paralog of this gene isCD209.
Specifications

Specifications

CLEC4M
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
IgG
-20°C
0.38 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit