Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Mouse anti-Human, Primate, Rhesus Macaque, Alexa Fluor™ 532, Clone: 14N8D7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227087AF532
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.Specifications
CLEC9a | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CLEC9A | |
A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. | |
Immunology, Stem Cells | |
283420 | |
Store at 4°C in the dark. |
Western Blot | |
14N8D7 | |
Western Blot | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
0.1 mL | |
Primary | |
Human, Primate, Rhesus Macaque | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title