Learn More
CLEC9a Mouse anti-Human, Primate, Rhesus Macaque, Alexa Fluor™ 594, Clone: 14N8D7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227087AF594
Catalog No. NP227087A94
Add to cart
Description
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Human,Primate,Rhesus Macaque samples. It is validated for Western Blot.
Specifications
Specifications
CLEC9a
14N8D7
Western Blot
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human, Primate, Rhesus Macaque
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CLEC9A
A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody.
0.1 mL
Primary
283420