Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC9a Antibody (14N8D7) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280673
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.Specifications
CLEC9a | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-3ug/ml | |
C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Orangutan, Chimp (100%), Canine, Panda, Equine (73%), Bovine (73%), Mouse (60%). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot | |
14N8D7 | |
Unconjugated | |
PBS | |
CLEC9A | |
A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. | |
0.1 mg | |
Immunology, Stem Cells | |
283420 | |
Human, Primate, Rhesus Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction