Learn More
CLGN Mouse anti-Human, HRP, Clone: 9C8G10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237395H
Catalog No. NBP237395H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CLGN
9C8G10
Western Blot, ELISA, Immunohistochemistry
Purified
CLGN
Purified recombinant fragment of human CLGN (AA: 249-405) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
calmegin
Mouse
IgG1
0.1 ml
Primary
1047