Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | CLIC2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180058
|
Novus Biologicals
NBP180058 |
100 μL |
Each of 1 for $436.00
|
|
Description
CLIC2 Polyclonal specifically detects CLIC2 in Human samples. It is validated for Western Blot.Specifications
CLIC2 | |
Polyclonal | |
Purified | |
RUO | |
NP_001280 | |
1193 | |
Synthetic peptide directed towards the C terminal of human CLIC2. Peptide sequence SAEEPPVSRRLFLDGDQLTLADCSLLPKLNIIKVAAKKYRDFDIPAEFSG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
chloride intracellular channel 2, chloride intracellular channel protein 2, XAP121CLIC2b | |
CLIC2 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title