Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Click N-Acetylmuramic acid - alkyne

Bacterial peptidoglycan derivative; suitable for 'click'-conjugation to fluorescent dyes

Manufacturer:  Tocris Bioscience™ 6798/2

Catalog No. 67-982

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Derivative of N-acetylmuramic acid (NAM) component of bacterial peptidoglycans. Incorporated into bacterial peptidoglycans during biosynthesis. Suitable for fluorescent labeling of peptidoglycans when 'click'-conjugated to a fluorescent dye.

Derivative of N-acetylmuramic acid (NAM) component of bacterial peptidoglycans. Incorporated into bacterial peptidoglycans during biosynthesis. Suitable for fluorescent labeling of peptidoglycans when 'click'-conjugated to a fluorescent dye.
Specifications

Specifications

97%
331.32
(2 R)-2-(((3 R,4 R,5 S,6 R)-2,5-dihydroxy-6-(hydroxymethyl)-3-(pent-4-ynamido)tetrahydro-2 H-pyran-4-yl)oxy)propanoic acid
2 mg
C14H21NO8
General Laboratory Research Reagent
2245794-65-2
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit