Learn More
FUJIFILM IRVINE SCIENTIFIC INC CLICKS MEDIUM 500ML DFS Item
Manufacturer:  FUJIFILM IRVINE SCIENTIFIC INC 9195500ML
Click's Medium is a modification of Eagle's Essential Medium with Hanks' Salts (HMEM) and contains higher concentrations of essential amino acids, in addition to non-essential amino acids, sodium pyruvate, and nucleic acid precursors.
Catalog No. NC0118954