Learn More
Vileda Professional™ ClickSpeed™ Disposable Mop Refill
Efficient, lightweight and simple to operate and use.
Manufacturer:  Vileda Professional™ 138653
Catalog No. 19-200-733
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • No pockets or velcro to trap dirt and bacteria
  • Easy mop preperation
  • Microfiber allows for excellent cleaning results
  • Lower water consumption and chemicals needed
  • For use with ClickSpeed™ Frame
  • Wash tested up to 300 cycles
  • Lightweight, allowing for higher mop loads per cycle, reducing washing costs considerably
Specifications
Specifications
Blue
50/Pk., 6 Pk./Cs.
Microfiber
Floor Mop
General Cleaning
Not Autoclavable
Non-sterile