Learn More
Invitrogen™ CLIF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114384
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human A549 whole cell, human HepG2 whole cell, rat ovary tissue, mouse lung tissue, mouse testis tissue. Flow: A431 cell, HL-60 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a basic helix-loop-helix transcription factor belonging to the PAS (PER, ARNT, SIM) superfamily. The PAS proteins play important roles in adaptation to low atmospheric and cellular oxygen levels, exposure to certain environmental pollutants, and diurnal oscillations in light and temperature. This protein forms a transcriptionally active heterodimer with the circadian CLOCK protein, the structurally related MOP4, and hypoxia-inducible factors, such as HIF1alpha. Consistent with its role as a biologically relevant partner of circadian and hypoxia factors, this protein is coexpressed in regions of the brain such as the thalamus, hypothalamus, and amygdala. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
Specifications
CLIF | |
Polyclonal | |
Unconjugated | |
ARNTL2 | |
4632430A05Rik; Arnt4; Arntl2; aryl hydrocarbon receptor nuclear translocator like 2; aryl hydrocarbon receptor nuclear translocator-like 2; aryl hydrocarbon receptor nuclear translocator-like protein 2; Basic-helix-loop-helix-PAS protein MOP9; BHLHE6; BMAL2; brain and muscle ARNT-like 2; brain and muscle Arnt-like protein 2 variant d; brain-muscle-ARNT-like protein 2; class E basic helix-loop-helix protein 6; CLIF; CYCLE-like factor; member of PAS protein 9; MGC149671; MGC149672; MOP9; PAS domain-containing protein 9; PASD9; transcription factor BMAL2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
272322, 362464, 56938 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Western Blot | |
500 μg/mL | |
PBS with 4 mg trehalose and 0.05 mg sodium azide | |
Q2VPD4, Q8WYA1 | |
ARNTL2 | |
E.coli-derived human ARNTL2 recombinant protein (Position: E461-T575). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.