Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Clindamycin hydrochloride, 20mg/mL in methanol, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
58207-19-5
C18H34Cl2N2O5S
461.439
MFCD07793327
AUODDLQVRAJAJM-NFOLQJLNSA-N
131632848
(2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
58207-19-5
C18H34Cl2N2O5S
461.439
MFCD07793327
AUODDLQVRAJAJM-NFOLQJLNSA-N
131632848
(2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
UN Number UN1230
Physical Form Liquid
Quantity 1mL
Chemical Name or Material Clindamycin hydrochloride, 20mg/mL in methanol, sterile-filtered

GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

RTECSNumber : GF2275000

storageNote1 : -30° to -10°C