Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin Hydrochloride, Monohydrate, USP, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
58207-19-5
C18H34Cl2N2O5S
461.44
AUODDLQVRAJAJM-UHFFFAOYNA-N
N-{2-chloro-1-[3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride
Cl.CCCC1CC(N(C)C1)C(=O)NC(C(C)Cl)C1OC(SC)C(O)C(O)C1O
58207-19-5
C18H34Cl2N2O5S
461.44
AUODDLQVRAJAJM-UHFFFAOYNA-N
N-{2-chloro-1-[3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl}-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride
Cl.CCCC1CC(N(C)C1)C(=O)NC(C(C)Cl)C1OC(SC)C(O)C(O)C1O

CL107, 58207-19-5, C18H33ClN2O5S•HCl•H2O

Spectrum™ Clindamycin Hydrochloride, Monohydrate, USP is an antibiotic used to treat infections with anaerobic bacteria. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 5 g
Packaging Amber Glass Bottle
CAS Max % 100%