Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Cliniqa™ LiniCAL™ Calibration Verifiers
Objectively verify instrument calibration and assess linearity for specified analytes
Manufacturer:  International Enzymes, Inc.Supplier Diversity Partner 98302
 View more versions of this product
Catalog No. 22-100-01
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Protein 2
14 Days Open Vial
Intended for use in the clinical laboratory to assess reagent or analyzer performance
5 x 1.5mL
Liquid
AAG, AAT, ASO, B2M, C3, C4, CER, CRP, FER, HPT, IgA, IgG, IgM, PAB, TRF
2° to 8°C