Learn More
Cliniqa™ LiniCAL™ Calibration Verifiers
Objectively verify instrument calibration and assess linearity for specified analytes
Manufacturer:  International Enzymes, Inc.Supplier Diversity Partner 98308
 View more versions of this product
Catalog No. 22-100-14
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Linearity Verifier
14 Days Open Vial
Intended for use in the clinical laboratory to assess reagent or analyzer performance
5 x 3mL
Liquid
ACTM, AMIK, CAFF, CAR, DIG, DISO, ETHO, GEN, LIDO, LI, NAPA, PHE, PHY, PRIM, PROC, QUIN, SALY, THE, TOB, VPA, VANC
2° to 8°C
Safety and Handling
ShelfLife : 2 Years