Learn More
Cliniqa™ LiniCAL™ Calibration Verifiers
Objectively verify instrument calibration and assess linearity for specified analytes
Manufacturer:  International Enzymes, Inc.Supplier Diversity Partner 98501
 View more versions of this product
Catalog No. 22-100-17
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Protein 1
30 Days Open Vial
Intended for use in the clinical laboratory to assess reagent or analyzer performance
6 x 1mL
Liquid
IgG, IgA, IgM, C3, C4, TRF
2° to 8°C