Cliniqa™ LiniCAL™ Calibration Verifiers

Objectively verify instrument calibration and assess linearity for specified analytes

Manufacturer: International Enzymes, Inc.Supplier Diversity Partner 98227

 View more versions of this product

Catalog No. 22-100-07

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Immunoprotein
    Format Liquid
    Stability 14 Days Open Vial
    Quantity 5 x 1mL,5 x 1mL
    Analytes ASO, IgA, IgG, IgM, TRF, PAB,ASO, IgA, IgG, IgM, TRF, PAB
    For Use With (Application) Intended for use in the clinical laboratory to assess reagent or analyzer performance
    Shelf Life 2 Years
    Storage Requirements 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit