Learn More
CLINT1 Mouse anti-Human, Clone: OTI10F12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806020
Catalog No. 50-166-7659
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.
Specifications
Specifications
CLINT1
Monoclonal
Unconjugated
CLINT1
Lyophilized
CLINT1
Human recombit protein fragment corresponding to amino acids 1-290 of human CLINT1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI10F12
PBS with 8% trehalose and no preservative; pH 7.3
Q14677
CLINT, ENTH, EPN4, EPNR
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9685