Learn More
Thermo Scientific™ ClipTip™ Filtered Pipette Tips (EU IVD/CE-marked) Available on GSA/VA Contract for Federal Government customers only.
Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Manufacturer:  Thermo Scientific™ 94420253
 View more versions of this product
Catalog No. 14-488-002
Add to cart
Description
Description
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Thermo Scientific™ E1-ClipTip, Thermo Scientific™ F1-ClipTip
ClipTip 50, filter, sterile
Filtered
Violet
5.1cm
Thermo Scientific™ E1-ClipTip, Thermo Scientific™ F1-ClipTip
Rack
50μL
Sterile
2.01 in.
ISO 9001, ISO 14001, ISO 13485
ClipTip
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.