Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLK1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen CLK1
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095935
View Documents
Novus Biologicals
NBP304429100UL
100 μg
Each for $320.71
Add to cart
 
NB095936
View Documents
Novus Biologicals
NBP30442920UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

CLK1 Polyclonal antibody specifically detects CLK1 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

CLK1
Unconjugated
Rabbit
Cell Cycle and Replication
PBS with 50% glycerol, pH7.3.
1195
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Human
CDC28/CDC2-like kinase, CDC-like kinase 1CLKEC 2.7.12.1, CLK/STY, dual specificity protein kinase CLK1, protein tyrosine kinase STY, STY
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human CLK1 (NP_004062.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit