Learn More
Abnova™ CLK4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057396T01
Catalog No. 89-939-868
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CLK4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.02kDa
Transient overexpression cell lysate was tested with anti-CLK4 antibody (Mfr. No. H00057396-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_020666.2
pCMV-CLK4 full-length
100 μL
Human