Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLM-9 (CD300c) Rat anti-Mouse, Unconjugated, Clone: ZAQ5, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CLM-9 (CD300c)
Applications ELISA, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT15111MI
View Documents
MilliporeSigma
MABT1511100UL
100 μL
Each for $372.00
Add to cart
 
MABT15112MI
View Documents
MilliporeSigma
MABT151125UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

CLM-9 (CD300c) Monoclonal antibody specifically detects CLM-9 (CD300c) in Mouse samples. It is validated for ELISA, Electron Microscopy, Immunohistochemistry (Paraffin).
Specifications

Specifications

CLM-9 (CD300c)
Unconjugated
Rat
CMRF35-like molecule 9;CD300 antigen-like family member G;Nepmucin;CD300g
Extracellular domain of mouse CMRF35-like molecule 9 (CLM-9; Nepmucin) protein fused with the Fc portion of human IgG
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunohistochemistry (Paraffin)
Purified
Mouse
Cd300lg, Clm9, D11Ertd736e
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit