CLMI Blueprint2 Safety Training Videos

Manufacturer: Comprehensive Loss Management, IncSupplier Diversity Partner STFRLCPD

 View more versions of this product

Catalog No. 19-124-127

  Check Availability
Add to Cart

    Specifications

    Includes Instructor guide
    Product Type Slips Trips and Falls - Retail
    Category Safety and Training Materials
    For Use With (Application) Covers mechanics and causes of ST and F, identification of hazards, avoiding hazards and preventative measures
    Format Video

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit